Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.09919

Supelco

Potassium hydroxide solution

c(KOH) = 0.5 mol/l (0.5 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

product name

Potassium hydroxide solution, for 1000 mlc(KOH) = 0.5 mol/l (0.5 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.5 M

technique(s)

titration: suitable

pH

>13.5 (20 °C in H2O)

density

1.58 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[K+].[O-][H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Impact of Modifications from Potassium Hydroxide on Porous Semi-IPN Hydrogel Properties and Its Application in Cultivation: This study discusses how potassium hydroxide modifications enhance the properties of semi-interpenetrating polymer network (semi-IPN) hydrogels, which are used in agricultural applications to improve soil water retention and plant growth (Tuan HNA et al., 2024).

  • Chitin nanofibrils assisted 3D printing all-chitin hydrogels for wound dressing: Potassium hydroxide plays a critical role in processing chitin nanofibrils for 3D printing hydrogels, which are then applied in medical fields as advanced wound dressing materials (Zheng Y et al., 2024).

  • Analytical study of KOH wet etch surface passivation for III-nitride micropillars: This research uses potassium hydroxide in wet etching processes to study its effects on surface passivation, crucial for enhancing the performance of III-nitride semiconductor micropillars in electronic applications (Seitz M et al., 2024).

  • Black goes green: single-step solvent exchange for sol-gel synthesis of carbon spherogels as high-performance supercapacitor electrodes: Potassium hydroxide is utilized to synthesize carbon spherogels via a green sol-gel process, demonstrating their efficacy as supercapacitor electrodes in energy storage technologies (Salihovic M et al., 2024).

  • Robust anion exchange membranes based on ionic liquid grafted chitosan/polyvinyl alcohol/quaternary ammonium functionalized silica for polymer electrolyte membrane fuel cells: This paper explores the application of potassium hydroxide in the development of durable anion exchange membranes for use in polymer electrolyte membrane fuel cells, highlighting its role in achieving high ionic conductivity (Jang HS et al., 2024).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service