Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09114

Supelco

Potassium hydroxide solution

c(KOH) = 0.5 mol/l (0.5 N), in ethanol, reag. Ph. Eur., ready-to-use volumetric solution for titration in non-aqueous media, Titriplex®

Synonym(s):

Potassium hydroxide ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352305
NACRES:
NA.21

Product Name

Potassium hydroxide solution in ethanol, c(KOH) = 0.5 mol/l (0.5 N), Titripur®, reag. Ph. Eur.

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

59 hPa ( 20 °C)

product line

Titripur®

form

liquid

autoignition temp.

425 °C (ethanol)

quality

Analyzed in our ISO 17025 accredited QC lab

expl. lim.

3.5-15.0 % (v/v) ethanol)

reaction suitability

reaction type: Acid-base reactions

concentration

0.5 M

technique(s)

titration: suitable

pH

14 (20 °C in H2O)

bp

78.3 °C/1013 hPa (ethanol)

mp

-114.5 °C (ethanol)

transition temp

flash point 12 °C (ethanol)

density

0.85 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Voltammetric Determination: Potassium hydroxide solution in ethanol is used in voltammetric determination of antioxidants in various matrices. A recent study demonstrated its application in the voltammetric determination of the antioxidant 2,6-di-tert-butyl-p-cresol (T501) in transformer oil using a graphite electrode. This method enhances the sensitivity and accuracy of antioxidant detection, crucial for ensuring the stability and performance of oils and other industrial fluids (Zhou et al., Analytical Letters, 2021).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.4975 - 0.5025 mol/L
Measurement uncertainty ± 0.0015 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 350 benzoic acid) by means of volumetric standard benzoic acid (article number 1.02401), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - (ethanol)

Flash Point(C)

12 °C - (ethanol)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service