Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09987

Supelco

Copper standard

1000 mg Cu, (CuCl₂ in H₂O) Titrisol®

Synonym(s):

Copper Reference Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Cu in H2O

technique(s)

AAS: suitable
ICP: suitable

pH

3.2 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Analysis Note

content (Cu): 1000 mg / Ampoule
Determination method: Complexometric titration.
(traceable to NIST - SRM 682)
Accuracy of the method: +/- 2mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service