Skip to Content
MilliporeSigma

1.09967

Aluminium standard

1000 mg Al, (AlCl3 in H2O) Titrisol®

Synonym(s):

Aluminium Reference Standard for ICP/AAS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

Titrisol®

form

concentrate

concentration

1000 mg Al in water

technique(s)

AAS: suitable, ICP: suitable

pH

2.5 (20 °C in H2O)

density

1.08 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Aluminium standard can be used as a calibration standard for the determination of aluminium content in water and beverage samples using graphite furnace atomic absorption spectrometry (GF-AAS)

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

1000 mg Al in H2O, prepared with high-purity AlCl3 and water.

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

target_organs

Lungs

supp_hazards

pictograms

Health hazardCorrosion

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library