Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38996

Supelco

Copper Standard for AAS

TraceCERT®, 1 g/L Cu in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Cu in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Leaching properties of coal ashes: Copper standards were critical in the analysis of leaching properties from coal ashes in a Malaysian power plant, ensuring precise calibration for atomic absorption spectroscopy, thus contributing significantly to environmental monitoring and research (Beddu et al., 2020).

  • Standard addition method in analytical chemistry: The application of rational functions for the standard addition method was explored, with copper AAS standards being crucial for establishing accurate analytical baselines, thus facilitating enhanced analytical outcomes in complex sample matrices (Gorazda et al., 2013).

Analysis Note

1 g/L Cu in 2% nitric acid, prepared with high purity Cu metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hukum Singh et al.
PloS one, 15(1), e0227380-e0227380 (2020-01-29)
The ever-increasing vehicle counts have resulted in a significant increase in air pollution impacting human and natural ecosystems including trees, and physical properties. Roadside plantations often act as a first defense line against the vehicular emissions to mitigate the impacts

Articles

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service