Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09436

Supelco

Buffer solution

(citric acid/sodium hydroxide) traceable to SRM from NIST and PTB pH 5.00 (20°C) Certipur®

Synonym(s):

Buffer solution pH 5.00 (20°C) (citric acid/sodium hydroxide)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161704
NACRES:
NA.25

grade

certified reference material

Quality Level

product line

Certipur®

pH

5.0 (25 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The buffer solution (citric acid/sodium hydroxide) is gravimetrically prepared using high-quality reagents in high-purity water. It is quality controlled and qualified in an ISO/IEC 17025 accredited calibration lab. Its pH value of 5.00 at 20°C is qualified using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266, IUPAC, NIST, USP, and Ph. Eur.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used in analytical procedures for the calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • Ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Long-lasting with a shelf life of 3 years
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an efficient audit and successful quality management system.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service