Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09407

Supelco

Buffer solution

(potassium dihydrogen phosphate/disodium hydrogen phosphate)traceable to SRM from NIST and PTB pH 7.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 7.00 (25°C) (potassium dihydrogen phosphate/disodium hydrogen phosphate)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161706
NACRES:
NA.25

grade

certified reference material

Quality Level

product line

Certipur®

pH

7.0 (25 °C in H2O)

bp

109 °C/1013 hPa

mp

-5 °C

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

A phosphate buffer, it is gravimetrically prepared using high-quality salts in high-purity water. The pH 7.00 of this potassium dihydrogen phosphate/disodium hydrogen phosphate is measured using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used in analytical procedures for the calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • Ready-to-use solution available in a sealed bottle and an innovative Titripac® packaging for a positive environmental impact due to reduced waste generation
  • Accompanied with a detailed certificate of analysis (CoA) for an effective audit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service