Skip to Content
MilliporeSigma

Skip To

1.02405

Sodium carbonate

volumetric standard, secondary reference material for acidimetry, traceable to NIST SRM Certipur®

Synonym(s):

anhydrous soda, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
UNSPSC Code:
41116107
NACRES:
NA.24
EC Index Number:
207-838-8
MDL number:
Beilstein/REAXYS Number:
4154566
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ISO 17025, reag. Ph. Eur., reag. USP

product line

Certipur®

form

solid

potency

4090 mg/kg LD50, oral (Rat)

technique(s)

titration: suitable

pH

11.16 (25 °C, 4 g/L in H2O)

mp

851 °C (lit.)

solubility

212.5 g/L

density

2.53 g/cm3 at 20 °C

bulk density

1100 kg/m3

storage temp.

15-25°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

General description

Sodium carbonate is a volumetric standard manufactured under stringent conditions and measured with the highest possible precision. Its mass fraction is determined by potentiometric titration, and it is qualified in a DIN EN ISO/IEC 17025 accredited laboratory.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website. Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used as a certified reference material (CRM) for the standardization of volumetric solutions in acidimetric titrations, according to USP and Ph. Eur.

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Features and Benefits

  • Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • Certified reference material according to ISO 17034
  • High-purity material traceable to NIST SRMs
  • Measured in a DIN EN ISO/IEC 17025 accredited laboratory
  • In accordance with the chapter reagents of USP and Ph. Eur
  • High-quality offering accurate titer determinations

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7136371345222321
description

volumetric standard, secondary reference material for acidimetry, traceable to NIST SRM Certipur®

description

reference material for titrimetry, certified by BAM, >99.5%

description

BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)

description

ACS reagent, anhydrous, ≥99.5%, powder or granules

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

-

technique(s)

-

product line

Certipur®

product line

-

product line

BioUltra

product line

-

Quality Level

300

Quality Level

100

Quality Level

200

Quality Level

200

potency

4090 mg/kg LD50, oral (Rat)

potency

-

potency

-

potency

-

pH

11.16 (25 °C, 4 g/L in H2O)

pH

-

pH

~12.0 (25 °C, 1 M in H2O)

pH

12 (25 °C, 106 g/L)

form

solid

form

-

form

powder or crystals

form

powder or granules


Still not finding the right product?

Explore all of our products under Sodium carbonate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters