Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.09911

Supelco

Hydrochloric acid

for the preparation of dilute volumetric solutions or for direct use c(HCl) = 5 mol/l (5 N) Combi-Titrisol

Synonym(s):

Hydrochloric acid solution, Hydrochloric acid 1.000, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

Hydrochloric acid solution, for the preparation of dilute volumetric solutions or for direct use c(HCl) = 5 mol/l (5 N) Combi-Titrisol

Agency

ISO 17025

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

1 M

technique(s)

titration: suitable

pH

<1 (20 °C in H2O)

density

1.1 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc.: This study explores the synthesis of zinc-ammonium phosphate from galvanic waste sludge, highlighting the use of hydrochloric acid solution in the extraction process, providing a sustainable approach for recycling industrial waste (Morgovan et al., 2024).

  • Corrosion inhibition of mild steel in hydrochloric acid solution by the expired Ampicillin drug.: This research investigates the effectiveness of expired Ampicillin in inhibiting the corrosion of mild steel in hydrochloric acid solution, offering a novel application for pharmaceutical waste in industrial settings (Alamry et al., 2023).

  • Contactless conductivity sensor employing moist paper as absorbent for in-situ detection of generated carbon dioxide gas.: This paper discusses the development of a contactless conductivity sensor using moist paper to detect carbon dioxide gas, utilizing hydrochloric acid solution in the analytical process for enhanced sensitivity and accuracy (Sonsa-Ard et al., 2020).

  • The microwave induced plasma with optical emission spectrometry (MIP-OES) in 23 elements determination in geological samples.: This study employs hydrochloric acid solution in sample preparation for the determination of 23 elements in geological samples using MIP-OES, demonstrating its effectiveness in analytical chemistry applications (Niedzielski et al., 2015).

  • Graphene sheets modified with polyindole for electro-chemical detection of dopamine.: The research highlights the use of hydrochloric acid solution in the preparation of polyindole-modified graphene sheets for the electrochemical detection of dopamine, showing significant improvements in sensor performance (Kumar and Prakash, 2014).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service