Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.04817

Supelco

Potassium chloride solution

3 mol/l, for storage and filling of electrodes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes
form:
liquid
Pricing and availability is not currently available.

Product Name

Potassium chloride solution, 3 mol/l

grade

for analytical purposes

Quality Level

form

liquid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

5.5 (20 °C in H2O)

density

1.13 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Surface Acoustic Wave Resonator Chip Setup for the Elimination of Interfering Conductivity Responses: This study introduces a novel application of potassium chloride solution in calibrating surface acoustic wave resonator chips, eliminating conductivity interferences in sensitive measurements. This technology is crucial for enhancing precision in both material science and biotechnological fields, offering robust analytical tools for researchers and developers (Rapp BE et al., 2024).

  • Development of a new continuous homogeneous liquid phase microextraction procedure: Highlighting the use of potassium chloride solution in developing new microextraction techniques for biochemical analysis, this study demonstrates the utility of potassium chloride in improving the detection of aliphatic amines in complex biological samples. This advancement supports the pharmaceutical industry and clinical research, promoting better analytical methods (Shagaghipour S et al., 2024).

  • Exploring the therapeutic potential of pomegranate juice for uterine relaxation: This research explores the biocompatibility and efficacy of potassium chloride solution used alongside natural agents for medical treatments, emphasizing its role in enhancing drug delivery and treatment strategies within the pharmaceutical sector. The study′s focus on non-invasive treatments showcases the potential for broader applications in healthcare (Al-Habib OAM et al., 2024).

  • A Simple Cost-Effective Method to Fabricate Single Nanochannels by Embedding Electrospun Polyethylene Oxide Nanofibers: Potassium chloride solution is used here to create precise nanochannels, a critical component in material science for developing advanced materials with specific ion transport properties. This research contributes significantly to the field of nanotechnology, enhancing the fabrication processes used in industrial applications (Zhou L et al., 2024).

  • Effects of Inorganic Salts on the Phase Separation of Partially Miscible Solutes: Investigating the effects of potassium chloride solution on phase separation processes, this study provides insights that are crucial for the development of new materials and the improvement of industrial separation techniques. The findings are especially relevant in environmental and chemical engineering, where precise control over phase behavior is essential (Zhao DY et al., 2024).

Analysis Note

Concentration c(KCl) = 3 mol/l
concentration: 2.9 - 3.1 mol/l

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service