Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.08816

Supelco

Zinc chloride

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Zinc chloride, Zinc dichloride, Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-592-0
NACRES:
NA.21
grade:
ACS reagent
form:
powder

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

1 mmHg ( 428 °C)
1.33 hPa ( 428 °C)

product line

EMSURE®

Assay

98.0-100.5% (ZnCl2, complexometric)

form

powder

potency

350 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤1.2% Oxide chloride (as ZnO)

pH

4.6-5.5 (10% in water)
5 (20 °C, 100 g/L in H2O)

bp

732 °C/1013 hPa

mp

287-304 °C
293 °C (lit.)

density

2.93 g/cm3 at 22 °C

bulk density

1400‑1800 kg/m3

anion traces

nitrate (NO3-): ≤0.003%
sulfate (SO42-): ≤0.002%

cation traces

Al:, passes test
Ca: ≤0.001%
Ca:, passes test
Cd: ≤0.0005%
Cu: ≤0.001%
Fe: ≤0.0005%
Fe:, passes test
K: ≤0.02%
Mg: ≤0.01%
Mg:, passes test
N: ≤0.002%
NH4+: ≤0.005%
Na: ≤0.005%
Pb: ≤0.001%
heavy metals:, passes test

storage temp.

2-30°C

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Influences of the Reaction Temperature and Catalysts on the Pyrolysis Product Distribution of Lignocellulosic Biomass (Aspen Wood and Rice Husk).: This research assesses the impact of reaction temperature and zinc chloride as a catalyst on the distribution of pyrolysis products from biomass sources. The findings are crucial for optimizing biofuel production processes, demonstrating how zinc chloride can alter product yields and characteristics (Sun et al., 2023).

  • Mechanistic study of ZnSe nanocrystal formation from zinc halides.: The study explores the mechanisms of zinc selenide (ZnSe) nanocrystal formation, particularly from zinc chloride precursors. This research is vital for applications in optoelectronics and photovoltaics, where precise control over nanocrystal properties is critical (Mingabudinova et al., 2023).

Linkage

Replaces: ZX0065-1; ZX0065-20; ZX0065

Analysis Note

Assay (complexometric, ZnCl₂): 98.0 - 100.5 %
Identity: passes test
Insoluble matter: ≤ 0.005 %
pH-value (10 %; water): 4.6 - 5.5
Nitrate (NO₃): ≤ 0.003 %
Oxide chloride (as ZnO): ≤ 1.20 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.002 %
AluminiumAluminium,Calcium,Magnesium,Heavy metals,Iron: passes test
Ca (Calcium): ≤ 0.001 %
Cd (Cadmium): ≤ 0.0005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.02 %
Mg (Magnesium): ≤ 0.01 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.005 %
Pb (Lead): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service