Skip to Content
MilliporeSigma

1.28221

Zinc chloride

EMPROVE® EXPERT, Ph. Eur., BP, USP

Synonym(s):

Zinc chloride, Dichlorozinc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
UNSPSC Code:
51472303
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur., USP

vapor pressure

1 mmHg ( 428 °C)

product line

EMPROVE® EXPERT

mol wt

136.30 g/mol

impurities

endotoxins, low

bp

293 °C

mp

293 °C (lit.)

solubility

851 g/L at 20 °C

density

2.93 g/cm3 at 22 °C

application(s)

liquid formulation
parenterals
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-25°C

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

General description

Zinc chloride is a mineral salt broadly used in pharmaceutical formulations, cosmetics and as supplement for nutrition.

Application

Mineral Salt

Packaging

1282211001 - 1kg in plastic bottle
1282218029 - 25kg in plastic bucket

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Zinc chloride


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library