Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

211273

Sigma-Aldrich

Zinc chloride

ACS reagent, ≥97%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥97%
grade:
ACS reagent
form:
crystalline powder
powder, crystals or chunks
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor pressure

1 mmHg ( 428 °C)

Assay

≥97%

form

crystalline powder
powder, crystals or chunks

reaction suitability

reagent type: catalyst
core: zinc

impurities

Oxychloride, passes test
≤0.005% insolubles

pH

5 (20 °C, 100 g/L)

mp

293 °C (lit.)

anion traces

nitrate (NO3-): ≤0.003%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.06%
Fe: ≤0.001%
K: ≤0.02%
Mg: ≤0.01%
NH4+: ≤0.005%
Na: ≤0.05%
Pb: ≤0.005%

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zinc chloride in combination with 1-ethyl-3-methylimidazolium chloride comprises a molten salt, which is employed in the electrodeposition studies of zinc on various substrates (glassy carbon and nickel).[1]

Application

Zinc chloride may be used as a catalyst for the Knoevenagel condensation of aromatic aldehydes with substituted acetonitriles to form carbonyl-methylene products under solvent-free conditions.[2] The activation of sugar cane bagasse with zinc chloride forms activated carbons for use as supercapacitor electrodes.[3]
ZnCl2 may be employed as a catalyst in the preparation of carbonyl-methylene condensation products[2] and poly(propylene fumarate).[4]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microstructure and electrochemical double-layer capacitance of carbon electrodes prepared by zinc chloride activation of sugar cane bagasse.
Rufford TE, et al.
Journal of Power Sources, 195(3), 912-918 (2010)
Albert K Shung et al.
Journal of biomaterials science. Polymer edition, 13(1), 95-108 (2002-05-11)
Diethyl fumarate and propylene glycol were reacted in the presence of a zinc chloride catalyst to synthesize poly(propylene fumarate) (PPF) over a period of 12 hours. The kinetics of the transesterification polymerization at 130 degrees C, 150 degrees C, and
Electrodeposition of zinc from a Lewis acidic zinc chloride-1-ethyl-3-methylimidazolium chloride molten salt.
Lin Y-F and Sun I.
Electrochimica Acta, 44(16), 2771-2777 (1999)
Zinc chloride as a new catalyst for Knoevenagel condensation.
Shanthan RP and Venkataratnam RV.
Tetrahedron Letters, 32(41), 5821-5822 (1991)
Manuella P Kaster et al.
Pharmacological reports : PR, 64(3), 706-713 (2012-07-21)
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service