Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08622

Supelco

3,3′,5,5′-Tetramethylbenzidine

GR for analysis

Synonym(s):

3,3′,5,5′-Tetramethylbenzidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
MDL number:
UNSPSC Code:
12352116
EC Index Number:
259-364-6
NACRES:
NA.05

Quality Level

form

solid

mp

166-170 °C

bulk density

430 kg/m3

storage temp.

15-25°C

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

Application


  • Effect of incubation time on nanozyme-catalyzed oxidation of 3,3′,5,5′-tetramethylbenzidine over silver nanoparticles.: This study investigates the influence of incubation time on the nanozyme-catalyzed oxidation of 3,3′,5,5′-tetramethylbenzidine (TMB) using silver nanoparticles. It highlights the kinetic parameters essential for optimizing the nanozyme activity and potential applications in biosensing and environmental monitoring (Hormozi Jangi et al., 2023).

  • Tetramethylbenzidine: An Acoustogenic Photoacoustic Probe for Reactive Oxygen Species Detection.: This research explores the use of TMB as a photoacoustic probe for detecting reactive oxygen species (ROS). It demonstrates TMB′s effectiveness in producing a photoacoustic signal upon reacting with various ROS, showcasing its potential in biological and medical diagnostics (Bresolí-Obach et al., 2020).

  • Investigation of vapochromism of 3,3′,5,5′-tetramethylbenzidine-tetrasilicicfluormica intercalation compounds through visible diffuse reflection and powder X-ray diffraction.: This paper examines the vapochromic properties of TMB intercalated in tetrasilicicfluormica using visible diffuse reflection and X-ray diffraction techniques. It reveals insights into the structural changes and potential applications in vapor sensing technologies (Kinoshita & Ishimaru, 2023).

  • The determination of the cholinesterase activity using 3,3′,5,5′-tetramethylbenzidine as an indicator.: This study introduces a new method for determining cholinesterase activity using TMB as an indicator. The method provides high sensitivity, reliability, and reproducibility, making it suitable for biochemical analyses and pharmaceutical applications (Koval′ska et al., 2021).

  • A facile photothermometric sensor of acid phosphatase based on CoOOH nanozymes-mediated 3,3′,5,5′-tetramethylbenzidine photothermal system.: This article presents a novel photothermometric sensor for acid phosphatase detection. It leverages CoOOH nanozymes and TMB to create a photothermal effect, providing a dual-mode detection system with applications in clinical diagnostics (Wei et al., 2021).

Analysis Note

Assay (perchloric acid titration): ≥ 99 %
Identity (IR-spectrum): conforms
Melting range: 166 - 170 °C
Loss on Drying (105°C): ≤ 0.2 %

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service