Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

861510

Sigma-Aldrich

3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate

97%

Synonym(s):

TMB dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H2(CH3)2-4-NH2]2·2HCl·xH2O
CAS Number:
Molecular Weight:
313.27 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

storage temp.

2-8°C

SMILES string

O.Cl.Cl.Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2

InChI

1S/C16H20N2.2ClH.H2O/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14;;;/h5-8H,17-18H2,1-4H3;2*1H;1H2

InChI key

KVCWTKDFVVSVSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Convenient water-soluble form of TMB for determination of peroxidase.
Noncarcinogenic substitute for benzidine.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service