Skip to Content
MilliporeSigma

Skip To

87748

3,3′,5,5′-Tetramethylbenzidine

TMB substrate, chromogenic, ≥98.0% (NT), powder

Synonym(s):

BM blue, Sure Blue TMB, TMB, TMB Blotting Plus, TMB substrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
[-C6H2(CH3)2-4-NH2]2
CAS Number:
Molecular Weight:
240.34
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
259-364-6
MDL number:
Beilstein/REAXYS Number:
2808541
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)

Quality Segment

assay

≥98.0% (NT)

form

powder

technique(s)

UV/Vis spectroscopy: suitable

mp

166-171 °C, 168-171 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2

InChI

1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3

InChI key

UAIUNKRWKOVEES-UHFFFAOYSA-N

Application

3,3′,5,5′-Tetramethylbenzidine (TMB) is a noncarcinogenic substitute (Ames test negative) for benzidine suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
3,3′,5,5′-Tetramethylbenzidine has been used as a substrate for peroxidase in:
  • enzyme-immunoassay (EIA)[1]
  • indirect competitive enzyme-linked immunosorbent assay (ELISA)[2]
  • sandwich enzyme-linked immunosorbent assay


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T288509743T5525
description

≥98.0% (NT)

description

≥98% (TLC)

description

standard for GC

description

tablet, 1 mg substrate per tablet

assay

≥98.0% (NT)

assay

≥98% (TLC)

assay

≥99.0% (GC)

assay

-

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

-

form

tablet

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

mp

166-171 °C, 168-171 °C (lit.)

mp

168-171 °C (lit.)

mp

166-171 °C, 168-171 °C (lit.)

mp

168-171 °C (lit.)


Still not finding the right product?

Explore all of our products under 3,3′,5,5′-Tetramethylbenzidine


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I Would Like to Kindly Ask for Stability Data for Temperature Excursion.

    1 answer
    1. This compound is relatively stable in powder form and ships at ambient temperatures globally, year round. Under these shipping conditions, transit times of up to 2 weeks are permitted. Brief temperature excursions are not expected to impact the quality of the product. However, extreme or long term temperature excursion studies have not been conducted. In the event of a temperature excursion, document the maximum temperature and duration, return the material to 2–8 °C, verify that the appearance remains white to light beige without caking or discoloration, and protect the product from light, and moisture. Please see the link below to access a Specification sheet. https://www.sigmaaldrich.com/specification-sheet/SIGMA/87748

      Helpful?

Reviews

No rating value

Active Filters