Skip to Content
MilliporeSigma
All Photos(1)

Documents

HX0635

Supelco

Hydrogen peroxide solution

30%, meets analytical specifications of USP/NF GR ACS

Synonym(s):

Hydrogen peroxide solution, Hydrogen Dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
51473503

product name

Hydrogen Peroxide Solution 30% W/W, meets analytical specifications of USP/NF GR ACS

Agency

meets analytical specifications of USP/NF GR ACS

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

form

liquid

shelf life

24 mo. (from mfg date)

concentration

29.0-32.0% (hydrogen peroxide)

impurities

≤0.0006 meq/g Titrable acid
≤0.5 ppm Iron (Fe)
≤1 ppm Heavy metals (as Pb)
≤2 ppm Chloride (Cl)
≤2 ppm Nitrate (NO3)
≤2 ppm Phosphate (PO4)
≤5 ppm Ammonium (NH4)
≤5 ppm Sulfate (SO4)

evapn. residue

≤0.002%

color

APHA: ≤10

shipped in

ambient

storage temp.

no temp limit

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Specifications

Tolerance: 29.0-32.0% W/W

Analysis Note

Expiration date: Exp 24 months from mfg date
Ammonium (NH4): 5ppm max
Assay: 29.0-32.0%
Chloride (Cl): 2ppm max
Color (APHA): 10 max
Evaporation residue: 0.002% max
Form: Clear liquid
Heavy metals (as Pb): 1ppm max
Iron (Fe): 0.5ppm max
Nitrate (NO3): 2ppm max
Phosphate (PO4): 2ppm max
Sulfate (SO4): 5ppm max
Titrable acid: 0.0006meq/g max

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen D Willis et al.
Molecular biology of the cell, 31(10), 1015-1031 (2020-03-12)
Environmental stress elicits well-orchestrated programs that either restore cellular homeostasis or induce cell death depending on the insult. Nutrient starvation triggers the autophagic pathway that requires the induction of several Autophagy (ATG) genes. Cyclin C-cyclin-dependent kinase (Cdk8) is a component
James P Zewe et al.
The Journal of cell biology, 219(3) (2020-03-27)
The polyphosphoinositides (PPIn) are central regulatory lipids that direct membrane function in eukaryotic cells. Understanding how their synthesis is regulated is crucial to revealing these lipids' role in health and disease. PPIn are derived from the major structural lipid, phosphatidylinositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service