Skip to Content
MilliporeSigma

1.08386

SAFC

Tris(hydroxymethyl)aminomethane

(Trometamol), EMPROVE® EXPERT, Ph. Eur., BP, ChP, JPC, USP

Pharma Manufacturing

Synonym(s):

Tromethamine, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-064-4

Agency

BP
ChP
JPC
Ph. Eur.
USP

Quality Level

vapor pressure

0.0267 hPa ( 20 °C)

product line

EMPROVE® EXPERT

form

solid

pH

10.2-10.6 (20 °C, 6 g/L in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)
219-220 °C/13.3 hPa

mp

167-172 °C (lit.)

solubility

678 g/L

density

1.35 g/cm3 at 23 °C

application(s)

ophthalmics
pharmaceutical

storage temp.

no temp limit

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Tris(hydroxymethyl)aminomethane (Trometamol) is a commonly used buffer component in biomolecule downstream chromatography steps as well as final liquid formulation.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service