Skip to Content
MilliporeSigma

1.37036

SAFC

di-Sodium hydrogen phosphate dihydrate

EMPROVE® EXPERT, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

di-Sodium hydrogen phosphate dihydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7

Agency

BP
Ph. Eur.
USP

Quality Level

description

EMPROVE® EXPERT Ph Eur,BP,USP

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

8.9-9.2 (5% in H2O)

mp

92.5 °C (Elimination of water of crystallisation)

solubility

93 g/L

density

2.1 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

di-Sodium hydrogen phosphate dihydrate is typically used as a phosphate buffer system component for biomolecule downstream chromatography steps or final liquid formulation.
Due to its low microbial and endotoxin limits, di-Sodium hydrogen phosphate dihydrate Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

The conjugation step in ADC production attaches a highly potent active pharmaceutical ingredient (HPAPI) to a mAb via a stable linker. Effective conjugation involves complex decisions, outlined on this page with services and products to assure success.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service