1.00314
Hydrochloric acid
37%, Emprove® ESSENTIAL, Ph. Eur., BP, JP, NF, fuming
Pharma Manufacturing
Synonym(s):
Hydrochloric acid, Hydrogen chloride solution
About This Item
Recommended Products
Product Name
Hydrochloric acid, fuming, 37%, EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF, ChP
Agency
BP
ChP
JP
NF
Ph. Eur.
Quality Level
vapor density
1.3 (vs air)
vapor pressure
190 hPa ( 20 °C)
3.23 psi ( 21.1 °C)
7.93 psi ( 37.7 °C)
product line
EMPROVE® ESSENTIAL
Assay
36.5-38.0% (alkalimetric)
form
liquid
concentration
37%
technique(s)
API processing | salt formation: suitable
pH
<1 (20 °C in H2O)
bp
>100 °C (lit.)
density
1.070 g/L at 20 °C
1.19 g/cm3 at 20 °C
application(s)
liquid formulation
ophthalmics
pharmaceutical
semi-solid formulation
storage temp.
2-25°C
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
Application
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Available by subscription or for a fee
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service