Skip to Content
MilliporeSigma

1.08421

SAFC

Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate)

EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP

Pharma Manufacturing

Synonym(s):

Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Beilstein:
3900609
UNSPSC Code:
12352107
EC Index Number:
205-358-3
Pricing and availability is not currently available.

Agency

BP
ChP
JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® ESSENTIAL

form

solid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

4-5 (20 °C, 50 g/L in H2O)

mp

248 °C (dec.) (lit.)

application(s)

liquid formulation
pharmaceutical

storage temp.

no temp limit

SMILES string

O.O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC(O)=O.OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2

InChI key

OVBJJZOQPCKUOR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Titriplex® III (EDTA, ethylene dinitrilo tetraacetic acid disodium salt dihydrate) is a commonly used chelating agent. As it is capable of forming complexes with divalent metal cations, it can inhibit diverse enzymes that require such cations for activity. It is broadly used as antioxidant in pharmaceutical formulations and cosmetics and can enhance the function of preservatives. The disodium salt provides improved solubility in water compared to standard EDTA.
Chelator of divalent cations. Inhibits enzymes, such as metalloproteases, that require divalent cations for activity.

Other Notes

Elimination of water of crystallization: 110 °C

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service