Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.08092

Supelco

Potassium antimony(III) oxide tartrate trihydrate

EMPLURA®

Synonym(s):

Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium tartrate, Potassium antimony tartrate, Tartar emetic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2(SbO)2C8H4O10 · 3 H2O
CAS Number:
Molecular Weight:
667.87
MDL number:
UNSPSC Code:
12352103
EC Index Number:
234-293-3
NACRES:
NA.21

Quality Level

product line

EMPLURA®

Assay

99.0-103.0% (iodometric)

form

solid

loss

≤ 2.7% loss on drying

pH

4.0 (20 °C, 50 g/L in H2O)

solubility

83 g/L

density

2.6 g/cm3 at 20 °C

bulk density

1250 kg/m3

cation traces

As: ≤0.015%
Pb: ≤0.002%

storage temp.

15-25°C

InChI

1S/2C4H4O6.2K.3H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;3*1H2;;/q2*-2;2*+1;;;;2*+3/p-4

InChI key

WBTCZEPSIIFINA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (iodometric): 99.0 - 103.0 %
Identity: passes test
As (Arsenic): ≤ 0.015 %
Pb (Lead): ≤ 0.002 %
Loss on drying: ≤ 2.7 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service