Skip to Content
MilliporeSigma
All Photos(2)

Documents

383376

Sigma-Aldrich

Potassium antimonyl tartrate trihydrate

ACS reagent, ≥99%

Synonym(s):

Antimony potassium tartrate trihydrate, Tartar emetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · 3H2O
CAS Number:
Molecular Weight:
667.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99%
99.0-103.0%

form

powder

impurities

≤0.020 meq/g Titr. acid or base

loss

≤2.7% loss on drying

mp

≥300 °C (lit.)

solubility

water: soluble

cation traces

As: ≤0.015%

SMILES string

O.O.O.[K+].[K+].O=C1O[Sb-]23OC1C4O[Sb-]5(OC(C(O2)C(=O)O3)C(=O)O5)OC4=O

InChI

1S/2C4H4O6.2K.3H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;3*1H2;;/q2*-2;2*+1;;;;2*+3/p-4

InChI key

WBTCZEPSIIFINA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium antimonyl tartrate trihydrate is a metalloid salt in which the antimony is in trivalent state (SbIII). Potassium antimonyl tartrate (PAT) is an antiparasitic agent predominantly used for treating infections caused by the parasite Leishmania. Its antiangiogenic and antitumor properties has been investigated in nonsmall-cell lung cancer (NSCLC) cells. Studies in Chinese hamster ovary cells show that it may prevent the repair of DNA double strand breaks (DSBs) caused by γ-irradiation.

Application

Potassium antimonyl tartrate trihydrate has been used in the dose-inhibition experiments to evaluate the dose-inhibition curves of antimony-based antileishmanial compounds against Leishmania promastigotes in medium 199.
Potassium antimonyl tartrate trihydrate may be used to prepare antimony (Sb(III)) stock solution in deionized water (100mg/L). It may also be employed in the spectrophotometric determination of total phosphorus in dry sediments.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nutrient contents in bottom sediment samples from a southern Brazilian microbasin.
Antunes M, et al.
Environmental earth sciences, 69(3), 959-968 (2013)
v
Vijayaraghavan K and Balasubramanian R.
Industrial & Engineering Chemistry Research, 50(17), 9864-9869 (2011)
Reduced Antimony Accumulation in ARM58-Overexpressing Leishmania infantum.
Schafer C, et al.
Antimicrobial Agents and Chemotherapy, 58(3), 1565-1574 (2014)
Valérie Lecureur et al.
International journal of oncology, 20(5), 1071-1076 (2002-04-17)
Potassium antimonyl tartrate (PAT), like arsenic trioxide (As2O3), has recently been shown to exert cytotoxicity towards acute promyelocytic leukemia (APL) cells. In the present study, we demonstrated that PAT treatment also inhibited cell growth of four acute myeloid leukemia (AML)
Sun-Hwa Nam et al.
Chemosphere, 75(7), 889-893 (2009-03-07)
Antimony is widespread in aquatic environment. Trivalent forms of antimony are known to be more toxic than other chemical species of antimony. In the present study, antimony potassium tartrate (APT), the trivalent inorganic forms of antimony, was selected as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service