Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.06559

Supelco

Sodium phosphate dibasic

anhydrous for analysis particle size about 0.2-1 mm (~18-80 mesh ASTM) EMSURE®

Synonym(s):

Sodium phosphate dibasic, di-Sodium hydrogen phosphate anhydrous, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7
NACRES:
NA.21
Assay:
≥99.0% (acidimetric)
grade:
anhydrous
form:
solid
solubility:
77 g/L

grade

anhydrous

Quality Level

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

loss

≤0.2% loss on drying, 105 °C

pH

8.7-9.3 (20 °C, 10 g/L in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

250 °C (decomposition)

solubility

77 g/L

bulk density

880 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
K: ≤0.01%
N: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 99.0 %
pH-value (5 %; water): 8.9 - 9.2
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Particle size (0.2 - 1 mm): about 90 %
Loss on Drying (105 °C): ≤ 0.2 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service