Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71640

Sigma-Aldrich

Sodium phosphate dibasic

puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.21
form:
powder (or crystals)

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (T)

form

powder (or crystals)

impurities

≤0.001% total nitrogen (N)

loss

≤0.2% loss on drying, 105 °C

pH

8.7-9.3 (25 °C, 50 mg/mL in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate dibasic is a sodium salt of phosphoric acid. It is highly hygroscopic and water-soluble compound used as a texturizer, emulsifier, and buffer. It can also be used as a reagent in water softeners, cleaners, and boiler treatments process.
Sodium phosphate is widely used buffer in molecular biology, biochemistry, and chromatographic studies. It exists in three forms: monobasic (NaH2PO4), dibasic (Na2HPO4), and tribasic (Na3PO4). Monobasic and dibasic forms in varying ratios are commonly used to compose the neutral sodium phosphate buffer solution.

Application

Sodium phosphate dibasic can be used:
  • As an efficient and low-cost catalyst for the synthesis of 2-aminochrome by the reaction between aromatic aldehyde, malononitrile, and α- or β-naphthol under solvent-free conditions.
  • As a buffer in the synthesis of cyclic, acyclic enones and α, β-unsaturated aldehydes from trimethylsilyl enol ethers by using palladium as a catalyst and oxone as oxidant.
  • As an efficient catalyst in the synthesis of 3,4-dihydropyrano[3,2-c] chromenes from aldehydes, malononitrile and 4-hydroxycoumarin.
  • As a phosphorous precursor with the phosphoric acid in the synthesis of hydroxyapatite by precipitation method using anionic surfactant SDS (sodium dodefcyl sulfate).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y C Chang et al.
Molecular and cellular biology, 14(7), 4912-4919 (1994-07-01)
Capsule formation plays a significant role in the pathogenicity of Cryptococcus neoformans. To study the molecular basis of capsule synthesis, the capsule-deficient phenotype of a mutant strain was complemented by transformation. A plasmid rescued from the resulting Cap+ transformant complemented
In vitro Studies: Inhibition of Nevirapine Metabolism by Nortriptyline in Hepatic Microsomes.
Usach I and PerisJE.
Bio-protocol, 5(19) (2015)
What is hemoglobin A1c? An analysis of glycated hemoglobins by electrospray ionization mass spectrometry.
Peterson KP, et al.
Clinical Chemistry, 44(9), 1951-1958 (1998)
Levon Manukyan et al.
Biomedicines, 8(4) (2020-04-01)
Coagulation Factor IX-rich protrhombin complex concentrate (FIX-PCC) is a therapeutic biologic product that consists of a mixture of several human plasma-derived proteins, useful for treating hemophilia B. Due to its complex composition, FIX-PCC is very challenging to bioprocess through virus
Kaisorn L Chaichana et al.
Journal of neuroscience methods, 180(1), 116-125 (2009-05-12)
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service