Skip to Content
MilliporeSigma

1.06586

Sodium phosphate dibasic

anhydrous for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium phosphate dibasic, di-Sodium hydrogen phosphate, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
NACRES:
NA.21
EC Index Number:
231-448-7
UNSPSC Code:
12352302
MDL number:
Assay:
≥99.0% (alkalimetric)
Grade:
ACS reagent
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

product line

EMSURE®

assay

≥99.0% (alkalimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

loss

≤ 0.2% loss on drying, 105 °C

pH

8.7-9.3 (20 °C, 10 g/L in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

>450 °C

bulk density

880 kg/m3

anion traces

chloride (Cl-): ≤0.002%, sulfate (SO42-): ≤0.005%

cation traces

Cu: ≤0.0003%, Fe: ≤0.001%, K: ≤0.01%, Pb: ≤0.001%, heavy metals: ≤0.001%

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Application

Sodium phosphate dibasic can be used as:
  • A component of the mobile phase for the analysis of guanidinoacetate methyltransferase using high-performance liquid chromatography (HPLC).
  • The source of phosphate ions in the synthesis of hydroxyapatite nanopowders.

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water, 25 °C): 8.7 - 9.3
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (ACS): ≤ 0.001 %
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.01 %
Pb (Lead): ≤ 0.001 %
Loss on Drying (105 °C): ≤ 0.2 %
Corresponds to ACS, Reag. Ph Eur

Other Notes

Replaces: SX0720; SX0720-1; SX0720-5; SX0720-7; SX0720-20

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Sodium phosphate dibasic


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library