Skip to Content
MilliporeSigma

Skip To

5.33005

Ammonium hydrogen carbonate

for LC-MS LiChropur

Synonym(s):

Ammonium hydrogen carbonate, Ammonium bicarbonate, Ammonium carbonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 g
Check Cart for Availability
$118.00

About This Item

Linear Formula:
(NH4)HCO3
CAS Number:
Molecular Weight:
79.06
UNSPSC Code:
12352301
EC Index Number:
213-911-5
NACRES:
NC.03
MDL number:

$118.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

67 hPa ( 20 °C)

product line

LiChropur

assay

99.0-101.0% (acidimetric)

form

solid

storage condition

(Tightly closed. Dry)

concentration

≤100% (Ammonium hydrogen carbonate)

technique(s)

HPLC: suitable

color

colorless

pH

8 (20 °C, 50 g/L in H2O)

mp

106 °C

solubility

220 g/L

application(s)

food and beverages

storage temp.

2-30°C

SMILES string

O=C(O)O.N

InChI

InChI=1S/CH2O3.H3N/c2-1(3)4;/h(H2,2,3,4);1H3, 1S/3CH2O3.2H3N.2Zn/c3*2-1(3)4;;;;/h3*(H2,2,3,4);2*1H3;;/q;;;;;2*+2/p-4

InChI key

ATRRKUHOCOJYRX-UHFFFAOYSA-N, QOPSSYZLVIENBR-UHFFFAOYSA-J

Application


  • Desorption of Ammonia Adsorbed on Prussian Blue Analogs by Washing with Saturated Ammonium Hydrogen Carbonate Solution.: This study explores the efficiency of ammonium hydrogen carbonate in removing adsorbed ammonia from Prussian Blue analogs, highlighting its potential application in environmental clean-up processes (Usuda et al., 2022).

  • Development and Validation of a Capillary Zone Electrophoresis-Tandem Mass Spectrometry Method for Simultaneous Quantification of Eight β-Lactam Antibiotics and Two β-Lactamase Inhibitors in Plasma Samples.: Although primarily focused on β-lactam antibiotics and β-lactamase inhibitors, this research utilized ammonium hydrogen carbonate in the sample preparation phase, showcasing its role in analytical methodologies (Cizmarova et al., 2024).

  • Development and validation of an LC-MS/MS method for quantifying diamorphine and its major metabolites 6-monoacetylmorphine, morphine, morphine-3-glucuronide, and morphine-6-glucuronide in human plasma.: This paper mentions the use of ammonium hydrogen carbonate in the process of drug metabolism analysis, indicating its utility in forensic and pharmacological testing (Thomann et al., 2024).

  • Fabrication of gelatin microspheres containing ammonium hydrogen carbonate for the tunable release of herbicide.: Discusses the innovative use of ammonium hydrogen carbonate in creating controlled-release gelatin microspheres for herbicide application, contributing to more sustainable agricultural practices (Liu et al., 2021).

  • GC-MS/MS Determination of Synthetic Cathinones: 4-chloromethcathinone, N-ethyl Pentedrone, and N-ethyl Hexedrone in Oral Fluid and Sweat of Consumers under Controlled Administration: Pilot Study.: While the focus is on synthetic cathinones, the role of ammonium hydrogen carbonate in sample preparation and stabilization in GC-MS/MS analyses reflects its importance in modern toxicological investigations (Nuñez-Montero et al., 2023).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.438351.01131A6141
assay

99.0-101.0% (acidimetric)

assay

-

assay

99.0-101.0% (acidimetric)

assay

≥99.0%

technique(s)

HPLC: suitable

technique(s)

-

technique(s)

-

technique(s)

-

concentration

≤100% (Ammonium hydrogen carbonate)

concentration

-

concentration

-

concentration

-

product line

LiChropur

product line

-

product line

EMPROVE® ESSENTIAL

product line

ReagentPlus®

form

solid

form

solid

form

solid

form

powder or crystals

application(s)

food and beverages

application(s)

-

application(s)

pharmaceutical
solid formulation

application(s)

-


Still not finding the right product?

Explore all of our products under Ammonium hydrogen carbonate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters