Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.03495

Sigma-Aldrich

1,1,2,2-Tetrachloroethane-D2

deuteration degree min. 99.5% for NMR spectroscopy MagniSolv

Synonym(s):

1,1,2,2-Tetrachloroethane-D2, Acetylene tetrachloride, 1,2-Dideutero-1,1,2,2-tetrachloroethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Cl4D2
CAS Number:
Molecular Weight:
169.86
MDL number:
UNSPSC Code:
12142201
EC Index Number:
251-634-1
NACRES:
NA.22

vapor pressure

6.6 hPa ( 20 °C)

Quality Level

form

liquid

technique(s)

NMR: suitable

impurities

≤0.02% Water (H<SUB>2</SUB>O) (NMR)

mp

-36 °C

solubility

3 g/L

density

1.595 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

ClC(Cl)(C(Cl)(Cl)[2H])[2H]

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H/i1D,2D

InChI key

QPFMBZIOSGYJDE-QDNHWIQGSA-N

Related Categories

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.5 %
Water (NMR, H₂O): ≤ 0.02 %
Performance test (NMR-spectrum): passes test

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service