The viscosity specification for the product is 100-120 mPas. The actual results for a specific lot or batch will be reported on the Certificate of Analysis.
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.21
UNSPSC Code:
41122605
Pricing and availability is not currently available.
Quality Segment
form
liquid
IVD
for in vitro diagnostic use
color
light yellow
refractive index
n20/D 1.515-1.517
density
1.02 g/cm3 at 20 °C
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
Immersion Oil - for Microscopy, is of an oily nature and has a defined refractive index of about 1,516. It is important that the refractive index (nD) is about 1.5, similar to that of glass. This enables a homogeneous oil immersion to be achieved. Immersion media are used in conjunction with immersion objectivs and are located between the surface of the specimen and the lens of the microscope. Immersion oils practically eliminate light beam deflection so that the effectiveness of the lens is considerably increased. Immersion Oil - for Microscopy is produced from a natural component and developed, certified and registered as CE / IVD product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
1046990001 is in drip plastic bottle and 1046990100 is 100ml in glass bottle.
1046990001 is in drip plastic bottle and 1046990100 is 100ml in glass bottle.
Analysis Note
Refractive index (n 20/D): 1.515 - 1.517
Density (d 20 °C/ 4 °C): 1.0245 - 1.0265
Transmission (380 nm; 1 cm): ≥ 65 %
Transmission (400 nm; 1 cm): ≥ 78 %
Transmission (450 nm; 1 cm): ≥ 90 %
Fluorescence (as quinine at 365 nm): ≤ 1500 ppb
Viscosity (20 °C): 100 - 120 mPa∙s
Suitability for microscopy: passes test
Density (d 20 °C/ 4 °C): 1.0245 - 1.0265
Transmission (380 nm; 1 cm): ≥ 65 %
Transmission (400 nm; 1 cm): ≥ 78 %
Transmission (450 nm; 1 cm): ≥ 90 %
Fluorescence (as quinine at 365 nm): ≤ 1500 ppb
Viscosity (20 °C): 100 - 120 mPa∙s
Suitability for microscopy: passes test
1 of 1
This Item | |||
|---|---|---|---|
| color light yellow | color yellow | color - | color - |
| form liquid | form - | form - | form liquid |
| application(s) diagnostic assay manufacturing | application(s) hematology | application(s) hematology | application(s) hematology |
| storage temp. 15-25°C | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| IVD for in vitro diagnostic use | IVD - | IVD - | IVD - |
| Quality Level 400 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
Still not finding the right product?
Explore all of our products under Immersion oil
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2
Storage Class
10 - Combustible liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




