Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358703

Sigma-Aldrich

1,1,2,2-Tetrachloroethane-d2

≥99.5 atom % D

Synonym(s):

1,2-Dideutero-1,1,2,2-tetrachloroethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CDCDCl2
CAS Number:
Molecular Weight:
169.86
Beilstein:
1699903
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.5 atom % D

Assay

≥99% (CP)

technique(s)

NMR: suitable

refractive index

n20/D 1.493 (lit.)

bp

145-146 °C/737 mmHg (lit.)

density

1.62 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

mass shift

M+2

SMILES string

[2H]C(Cl)(Cl)C([2H])(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H/i1D,2D

InChI key

QPFMBZIOSGYJDE-QDNHWIQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1,2,2-Tetrachloroethane-d2 is 1,1,2,2-Tetrachloroethane in which all hydrogen atoms are replaced by deuterium. It is commonly employed as an NMR solvent.

Application

1,1,2,2-Tetrachloroethane-d2 is used as a solvent:
  • In the synthesis of cyclic olefin copolymer foam (COC), which is a super thermal insulation material in energy-management applications.
  • In the NMR measurement to determine various intermediate compounds.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E London et al.
Science advances, 5(5), eaav2336-eaav2336 (2019-05-30)
Interest in high-spin organic materials is driven by opportunities to enable far-reaching fundamental science and develop technologies that integrate light element spin, magnetic, and quantum functionalities. Although extensively studied, the intrinsic instability of these materials complicates synthesis and precludes an
Randi Zhang et al.
Dalton transactions (Cambridge, England : 2003), 48(47), 17488-17498 (2019-11-21)
Five structurally related bis(arylimino)pyridine-iron(ii) chloride complexes, [2-[CMeN{2,6-{(4-FC6H4)2CH}2-4-NO2}]-6-(CMeNAr)C5H3N]FeCl2 (Ar = 2,6-Me2C6H3Fe1, 2,6-Et2C6H3Fe2, 2,6-i-Pr2C6H3Fe3, 2,4,6-Me3C6H2Fe4, and 2,6-Et2-4-MeC6H2Fe5), incorporating one N-2,6-bis{di(4-fluorophenyl)methyl}-4-nitrophenyl group and one distinct N-aryl group, have been prepared in good yield through the interaction of the corresponding free ligands (L1-L5) with
Haobing Wang et al.
Journal of the American Chemical Society, 141(32), 12624-12633 (2019-07-26)
The precise control of regio- and stereochemistry in the cyclopolymerization of heteroatom-functionalized α,ω-dienes is of much interest and importance, but has remained a challenge to date. We report herein the regio-, diastereoselective and stereoregular cyclopolymerization of ether- and thioether-functionalized 1,6-heptadienes
Liwei Guo et al.
Dalton transactions (Cambridge, England : 2003), 48(17), 5604-5613 (2019-04-09)
A novel family of 2,6-bis[1-(p-dibenzocycloheptylarylimino)ethyl]pyridylcobalt dichlorides Co1-Co4 were synthesized and fully characterized by FT-IR, 1H NMR, and elemental analysis as well as X-ray diffraction analysis. The 1H NMR spectra of these paramagnetic cobalt complexes Co1-Co3 showed similar spectra with one
Emilie Steiner et al.
Magnetic resonance in chemistry : MRC, 53(8), 596-602 (2015-06-23)
Host-guest complexes between cryptophane-A analogue with butoxy groups (cryptophane-But) and chloromethanes (chloroform, dichloromethane) were investigated in the solid state by means of magic-angle spinning (13)C NMR spectroscopy. The separated local fields method with (13)C-(1)H dipolar recoupling was used to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service