Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.05362

Millipore

L-Isoleucine

for biochemistry

Synonym(s):

L-Isoleucine, iso-Leucine, Ile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-798-2
NACRES:
NA.21

Quality Level

form

solid

pH

5.5-6.5 (20 °C, 40 g/L in H2O)

mp

279-280 °C (decomposition)

solubility

40 g/L

bulk density

470 kg/m3

storage temp.

2-30°C

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1

InChI key

AGPKZVBTJJNPAG-WHFBIAKZSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, crystalline powder or flakes
Appearance of solution (1 mol/l, hydrochloric acid 1 mol/l): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +36.2 to +37.2
UV-Absorption (260 nm, 1 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.10
UV-Absorption (280 nm, 1 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.10
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.5 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service