Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02692

Supelco

Celite® 503 Diatomaceous Earth

Celite® 503. CAS 68855-54-9, pH 10 (100 g/l, H₂O, 20 °C).

Synonym(s):

Celite® 503

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
EC Index Number:
272-489-0
NACRES:
NB.21

product name

Celite® 503, Celite® 503. CAS 68855-54-9, pH 10 (100 g/l, H₂O, 20 °C).

Quality Level

technique(s)

HPLC: suitable

pH

10 (20 °C, 100 g/L in H2O)

mp

>450 °C

solubility

0.0037 g/L

density

2.36 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

15-25°C

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Application

Celite® 503. CAS 68855-54-9, pH 10 (100 g/l, H₂O, 20 °C).

Analysis Note

Identity: passes test
Loss on ignition (600 °C): ≤ 0.5 %
Particle size (< 0.1 mm): ≥ 85 %

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service