Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02039

Supelco

Cesium chloride

99.995 Suprapur®

Synonym(s):

Cesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClCs
CAS Number:
Molecular Weight:
168.36
UNSPSC Code:
12352302
EC Index Number:
231-600-2
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

Assay

99.995%

form

solid

potency

2004 mg/kg LD50, oral (Rat)

pH

7-9 (20 °C, 50 g/L in H2O)

mp

646 °C

solubility

1860 g/L

density

3.97 g/cm3 at 20 °C

bulk density

1800 kg/m3

storage temp.

2-30°C

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Related Categories

Application


  • Optimizing high-throughput viral vector characterization with density gradient equilibrium analytical ultracentrifugation.: This study utilizes cesium chloride for optimizing high-throughput characterization of viral vectors using density gradient equilibrium analytical ultracentrifugation, enhancing the efficiency and accuracy of viral vector analysis. (Sternisha et al., 2023).

  • Mammalian Reoviruses: Propagation, Quantification, and Storage.: Cesium chloride is used in this study to facilitate the propagation, quantification, and storage of mammalian reoviruses, contributing to better handling and study of these viruses in research settings. (Coombs, 2023).

  • Large-scale purification of functional AAV particles packaging the full genome using short-term ultracentrifugation with a zonal rotor.: This paper discusses the large-scale purification of adeno-associated virus particles using cesium chloride in ultracentrifugation, which is crucial for gene therapy applications. (Wada et al., 2023).

Analysis Note

Purity (metallic): ≥ 99.995 %
Assay (argentometric): ≥ 99.5 %
Specific absorbance (260 nm; 50 %; 1 cm; water): ≤ 0.020
Specific absorbance (280 nm; 50 %; 1 cm; water): ≤ 0.020
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 20 ppm
Total nitrogen (N): ≤ 10 ppm
Al (Aluminium): ≤ 0.05 ppm
B (Boron): ≤ 0.05 ppm
Ba (Barium): ≤ 2.0 ppm
Ca (Calcium): ≤ 0.50 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.005 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.005 ppm
Fe (Iron): ≤ 0.020 ppm
K (Potassium): ≤ 2 ppm
La (Lanthanum): ≤ 0.005 ppm
Li (Lithium): ≤ 0.2 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Na (Sodium): ≤ 4.0 ppm
Ni (Nickel): ≤ 0.020 ppm
Pb (Lead): ≤ 0.030 ppm
Rb (Rubidium): ≤ 5 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.20 ppm
Tl (Thallium): ≤ 0.20 ppm
Y (Yttrium): ≤ 0.005 ppm
Yb (Ytterbium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.020 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service