Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219650

Millipore

Cesium Chloride, Molecular Biology Grade

Used for density gradient centrifugation. Commonly used for fractionation of nucleic acids, ribosomal subunits, proteins, glycoproteins, and viruses.

Synonym(s):

Cesium Chloride, Molecular Biology Grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClCs
CAS Number:
Molecular Weight:
168.36
UNSPSC Code:
12352302
NACRES:
NA.21

grade

Molecular Biology

Quality Level

Assay

≥99% (silver nitrate titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 500 mg/mL

cation traces

Pb: ≤0.01%

foreign activity

DNases, RNases, proteases, none detected

shipped in

ambient

storage temp.

10-30°C

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

General description

Used for density gradient centrifugation. Commonly used for fractionation of nucleic acids, ribosomal subunits, proteins, glycoproteins, and viruses. Absorbance: (3 M, H2O, 260 nm): ≤0.1; (3 M, H2O, 280 nm): ≤0.02.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service