Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02041

Supelco

Cesium chloride

EMPLURA®

Synonym(s):

Cesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClCs
CAS Number:
Molecular Weight:
168.36
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-600-2
NACRES:
NA.21
Assay:
≥99.0% (silver nitrate titration)
form:
solid
solubility:
1860 g/L

Quality Level

product line

EMPLURA®

Assay

≥99.0% (silver nitrate titration)

form

solid

potency

2004 mg/kg LD50, oral (Rat)

pH

7-9 (20 °C, 50 g/L in H2O)

mp

646 °C

solubility

1860 g/L

density

3.97 g/cm3 at 20 °C

bulk density

1800 kg/m3

anion traces

sulfate (SO42-): ≤0.03%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
K: ≤0.005%
Mg: ≤0.002%
Na: ≤0.005%
Sr: ≤0.002%

storage temp.

2-30°C

SMILES string

[Cs+].[Cl-]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 99.0 %
Sulfate (SO₄): ≤ 0.03 %
Ba (Barium): ≤ 0.002 %
Ca (Calcium): ≤ 0.002 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.002 %
Na (Sodium): ≤ 0.005 %
Sr (Strontium): ≤ 0.002 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service