Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.01810

Supelco

Conductivity water ( nominal 0 mS/cm)

Test solution for measurement of electrolytic conductivity, traceable to PTB Certipur®

Synonym(s):

Water, Deionized water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

vapor pressure

23 hPa ( 20 °C)

Quality Level

form

liquid

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

15-25°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The nominal conductivity value of the standard is determined by an accredited Merck calibration laboratory for pH value and electrolytic conductivity.

This product is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany.

Application

Used as a control sample to measure conductivity of aqueous electrolyte solutions.

Analysis Note

Electrical conductivity (25 °C): 0 - 0.002 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service