Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.01811

Supelco

Potassium chloride solution

nominal 0.015 mS/cm, reference material for the measurement of electrolytic conductivity, traceable to PTB (c=0.0001 mol/l) Certipur®

Synonym(s):

Potassium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

product name

Potassium chloride solution Certified Reference Material, nominal 0.015 mS/cm, Certified Reference Material for the measurement of electrolytic conductivity, traceable to PTB (c=0.0001 mol/l) Certipur®

grade

reference material

Quality Level

product line

Certipur®

application(s)

environmental

format

single component solution

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany.

The nominal conductivity value of the standard is determined using a conductivity meter and calibrated with primary reference material following DIN EN ISO / IEC 17025.

Application

Intended for use in aqueous solution as a calibration standard for the measurement of the conductivity cell constant or as a control sample.

Analysis Note

Electrical conductivity (25 °C): 0.013 - 0.017 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service