Skip to Content
MilliporeSigma
All Photos(1)

Documents

COND25

Supelco

Conductivity Standard - 25 μmhos/cm

certified reference material

Synonym(s):

25 μmhos/cm Conductivity Calibration Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

100
300

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-25-0.500

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This ready-to-use certified reference material (CRM) is produced and qualified following ISO 17034 and ISO/IEC 17025. It is manufactured gravimetrically with traceability to SI and higher order standards from National Institute of Standards and Technology (NIST). Its certified value is based on purity determination by mass balance and gravimetrically prepared value. It is delivered in a 500 mL HDPE bottle.

All information regarding metrological traceability, uncertainty, homogeneity and stability is documented in the certificate of analysis (CoA).

Application

Suitable for use in calibration of instruments for conductivity measurements.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service