Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.01553

Supelco

Potassium chloride solution

nominal 1.41 mS/cm, reference material for the measurement of electrolytic conductivity, traceable to PTB and NIST (c=0.01 mol/l) Certipur®

Synonym(s):

Potassium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

product name

Potassium chloride solution Certified Reference Material, nominal 1.41 mS/cm, Certified Reference Material for the measurement of electrolytic conductivity, traceable to PTB (c=0.01 mol/l) Certipur®

grade

reference material

Quality Level

product line

Certipur®

pH

7 (20 °C in H2O)

bp

100 °C/1013 hPa

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (CRM) is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany.

Application

Standard for use in measurement of electrolytic conductivity.

Analysis Note

Electrical conductivity (25 °C): 1.39 - 1.43 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service