Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-041

Supelco

Vitamin K1-13C6 solution

5 μg/mL in ethanol, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

Vitamin K1 Carbon-13 Labeled Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2513C6H46O2
Molecular Weight:
456.65
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5 μg/mL in ethanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O=C1c2c(cccc2)C(=O)C(=C1C\C=C(\CCC[C@@H](CCC[C@@H](CCCC(C)C)C)C)/C)C

InChI

1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1

InChI key

MBWXNTAXLNYFJB-NKFFZRIASA-N

General description

This stable-labeled internal standard is suitable for quantification of Vitamin K1 in LC-MS/MS clinical testing applications such as assessment of vitamin K deficiency as well as food and nutrition testing applications. Vitamin K1 (phylloquinone) is a fat-soluble vitamin that serves as a cofactor for vitamin K-dependent (VKD) proteins and enzymes which activate blood-clotting factors. Vitamin K1 is critical to many physiological processes including bone metabolism, arterial calcification, growth control, and signal transduction.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service