Skip to Content
MilliporeSigma

Skip To

V-031

Vitamin K2 (MK-4) solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H40O2
CAS Number:
Molecular Weight:
444.65
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

mp

-45.69 °C (at 1013 hPa)

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CCC=C(C)C)=C(C)C(C2=CC=CC=C21)=O

InChI

1S/C31H40O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,12,14,16,18-20H,9-11,13,15,17,21H2,1-6H3/b23-14+,24-16+,25-20+

InChI key

DKHGMERMDICWDU-GHDNBGIDSA-N

General description

Snap-N-Spike®
Snap-N-Shoot®
Vitamin K2 (MK-4) or menaquinone-4 is an essential cofactor for enzymes that activate blood-clotting factors as well as vitamin K-dependent (VKD) proteins involved in bone metabolism, arterial calcification, growth control, and signal transduction.

Application

This certified solution standard is suitable for use as starting material in the preparation of linearity standards, calibrators, or controls in LC-MS/MS vitamin K2 testing applications such as assessment of vitamin K2 deficiency and research of VKD proteins.
Vitamin K2 (MK-4) also known as menaquinone may be used as a certified reference material (CRM) for the determination of the analyte in biological and animal products by high-performance liquid chromatography (HPLC) method.

Legal Information

Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V-030V-042V-041
application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters