Skip to Content
MilliporeSigma
All Photos(1)

Documents

S-037

Supelco

JWH-073 N-(3-hydroxybutyl) metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

JWH-073 3-Hydroxybutyl metabolite solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21NO2
Molecular Weight:
343.42
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(O)CCn1cc(C(=O)c2cccc3ccccc23)c4ccccc14

InChI

1S/C23H21NO2/c1-16(25)13-14-24-15-21(19-10-4-5-12-22(19)24)23(26)20-11-6-8-17-7-2-3-9-18(17)20/h2-12,15-16,25H,13-14H2,1H3

InChI key

YPRJGJFJMZOUSH-UHFFFAOYSA-N

General description

The major and most prominent urinary metabolite of JWH-073, a synthetic cannabinoid found in several versions of the herbal mixture Spice. Our certified Spice cannabinoid solution standards are DEA-exempt allowing laboratories to order these standards without DEA paperwork.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This study demonstrates the impact of various mobile phase modifiers on the separation; formate modifiers outperform acetate in terms of MS signals (or sensitivity) and chromatographic resolution.

Chromatograms

application for HPLCapplication for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service