Skip to Content
MilliporeSigma
All Photos(1)

Documents

S-041

Supelco

Spice Cannabinoid Mix 2 solution

100 μg/mL each component in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

AM2201
JWH-019
JWH-081
JWH-122

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland)

concentration

100 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A certified multi-component solution mix of Spice cannabinoids JWH-019, JWH-122, JWH-081, and AM-2201. Cases of abuse for these four Spice cannabinoids, similar in structure to JWH-018 and JWH-073, have been observed across the US from California and Texas to Pennsylvania and Missouri. This certified solution mix is suitable for GC/MS and LC/MS applications in forensic analysis, clinical toxicology, and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service