Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
grade
certified reference material
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
O=C(C1=CC=CC2=C1C=CC=C2)C3=CN(CCCCC(O)=O)C4=C3C=CC=C4
InChI
1S/C24H21NO3/c26-23(27)14-5-6-15-25-16-21(19-11-3-4-13-22(19)25)24(28)20-12-7-9-17-8-1-2-10-18(17)20/h1-4,7-13,16H,5-6,14-15H2,(H,26,27)
InChI key
BYIVGHIYNFQIJX-UHFFFAOYSA-N
General description
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant® | description 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant® | description 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant® | description 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant® |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| format single component solution | format single component solution | format single component solution | format single component solution |
| form liquid | form liquid | form liquid | form liquid |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






