Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IMPM-005-06

Supelco

10-Oxomorphine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

10-Ketomorphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO4
CAS Number:
Molecular Weight:
299.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C(=O)[C@@H]1[C@@H]2C=C[C@@H]4O)c35

InChI

1S/C17H17NO4/c1-18-7-6-17-9-3-5-11(20)16(17)22-15-10(19)4-2-8(12(15)17)14(21)13(9)18/h2-5,9,11,13,16,19-20H,6-7H2,1H3/t9-,11-,13-,16-,17-/m0/s1

InChI key

HNJBQZPKNKFLFM-CBEJNMEVSA-N

General description

A certified solution standard of the decomposition product measured in manufactured morphine sulfate drug products or APIs using LC or LC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Proksa
Die Pharmazie, 39(10), 687-688 (1984-10-01)
10-Oxomorphine, a decomposition product of morphine, was synthesized from 6-O-acetylcodeine, which was oxidized with chromic acid to 6-O-acetyl-10-hydroxycodeine. This substance was then oxidized with manganese dioxide to 6-O-acetyl-10-oxocodeine, which on hydrolysis and O-demethylation gave 10-oxomorphine. UV-, fluorimetric spectrometry, TLC and
Seán S Kelly et al.
Journal of pharmaceutical sciences, 92(3), 485-493 (2003-02-15)
Stability testing of morphine sulfate formulated with nonpareil sugar seeds (consisting of sucrose and starch) and fumaric acid revealed the formation of the three impurities 5-(hydroxymethyl)-2-furfural, 10-hydroxymorphine and 10-oxomorphine. 5-(Hydroxymethyl)-2-furfural was isolated via semipreparative HPLC utilizing volatile mobile phase constituents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service