Skip to Content
MilliporeSigma
All Photos(1)

Documents

IMPM-005-05

Supelco

10-Hydroxymorphine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO4
CAS Number:
Molecular Weight:
301.34
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C(O)[C@@H]1[C@@H]2C=C[C@@H]4O)c35

InChI

1S/C17H19NO4/c1-18-7-6-17-9-3-5-11(20)16(17)22-15-10(19)4-2-8(12(15)17)14(21)13(9)18/h2-5,9,11,13-14,16,19-21H,6-7H2,1H3/t9-,11-,13-,14?,16-,17-/m0/s1

InChI key

ISVKWTNAIRJURI-AKUCRQHZSA-N

General description

10-Hydroxymorphine is a potential impurity from the production of morphine pharmaceutical formulations. Morphine is a potent opiate analgesic drug used to relieve both acute and chronic severe pain. This Certified Snap-N-Spike® Solution is applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as impurity identification.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Proksa
Journal of pharmaceutical and biomedical analysis, 20(1-2), 179-183 (2000-03-07)
Separation of morphine and its oxidation products (10-S-hydroxymorphine, pseudomorphine and morphine N-oxide) by capillary zone electrophoresis in Tris-borate buffer in the presence of cyclodextrins was studied. Pyridoxine was used as an internal standard.
Morphine impurity with opioid activity is identified as 10 alpha-hydroxymorphine.
H Farsam et al.
Pharmaceutical research, 7(11), 1205-1207 (1990-11-01)
Seán S Kelly et al.
Journal of pharmaceutical sciences, 92(3), 485-493 (2003-02-15)
Stability testing of morphine sulfate formulated with nonpareil sugar seeds (consisting of sucrose and starch) and fumaric acid revealed the formation of the three impurities 5-(hydroxymethyl)-2-furfural, 10-hydroxymorphine and 10-oxomorphine. 5-(Hydroxymethyl)-2-furfural was isolated via semipreparative HPLC utilizing volatile mobile phase constituents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service