Skip to Content
MilliporeSigma

P-103

Posaconazole solution

2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H42F2N8O4
CAS Number:
Molecular Weight:
700.78
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
200-659-6
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

2.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

room temp

SMILES string

Fc1c(ccc(c1)F)[C@@]3(OC[C@H](C3)COc4ccc(cc4)N5CCN(CC5)c6ccc(cc6)N7C=NN(C7=O)[C@H]([C@@H](O)C)CC)C[n]2ncnc2

InChI

1S/C37H42F2N8O4/c1-3-35(26(2)48)47-36(49)46(25-42-47)31-7-5-29(6-8-31)43-14-16-44(17-15-43)30-9-11-32(12-10-30)50-20-27-19-37(51-21-27,22-45-24-40-23-41-45)33-13-4-28(38)18-34(33)39/h4-13,18,23-27,35,48H,3,14-17,19-22H2,1-2H3/t26-,27+,35-,37-/m0/s1

InChI key

RAGOYPUPXAKGKH-XAKZXMRKSA-N

General description

A certified Snap-N-Spike® Solution suitable as starting material for calibrators, controls, or linearity standards in posaconazol therapeutic drug monitoring or clinical and diagnostic testing applications in patient blood, serum, or plasma samples by LC-MS/MS or HPLC. Sold under trade names Noxafil® and Posanol, posaconazole is a triazole antifungal drug prescribed for treatment of invasive fungal infections in severely immunocompromised patients.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Noxafil is a registered trademark of MSD
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

49.5 °F

Flash Point (°C)

9.7 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library