Skip to Content
MilliporeSigma
All Photos(1)

Documents

M-006

Supelco

Morphine-D3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16NO3D3
CAS Number:
Molecular Weight:
288.36
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@@H](C=C1)[C@H]2C(C1C3C4)(CCN3C([2H])([2H])[2H])C5=C4C=CC(O)=C5O2

InChI

1S/C17H19NO3/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18/h2-5,10-11,13,16,19-20H,6-8H2,1H3/t10-,11+,13-,16-,17-/m0/s1/i1D3

InChI key

BQJCRHHNABKAKU-LLQNGXJRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This stable-labeled internal standard is suitable for quantiation of morphine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, or isotope dilution methods. Morphine is a very potent opiate analgesic drug used to relieve both acute and chronic severe pain. Sold under trade names including MS Contin®, Kadian®, Oramorph®, and Roxanol, morphine is also a commonly abused recreational drug due to its widespread availability and high potential for addiction.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Kadian is a registered trademark of Actavis Elizabeth LLC
MS Contin is a registered trademark of Purdue Frederick Company
Oramorph is a registered trademark of AAIPharma, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service