Skip to Content
MilliporeSigma
All Photos(2)

Documents

900693

Sigma-Aldrich

9-Mesityl-1,3,6,8-tetramethoxy-10-phenylacridin-10-ium tetrafluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32NO4BF4
CAS Number:
Molecular Weight:
581.41
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

core: acridinium
reagent type: catalyst
reaction type: Photocatalysis

photocatalyst activation

450 nm

SMILES string

CC1=CC(C)=CC(C)=C1C2=C(C(OC)=CC(OC)=C3)C3=[N+](C4=CC=CC=C4)C5=CC(OC)=CC(OC)=C52.FB(F)F.[F-]

Application

Robust acridinium-based photocatalyst reported by Nicewicz and coworkers as an alternative to transition-metal-based photocatalysts with higher chemical stability and attenuated redox potential.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Nicewicz lab is focused on the discovery of new and powerful reaction methodologies that proceed via the intermediacy of highly reactive cation radical species. Included in these transformations are anti-Markovnikov selective additions of amines, alcohols, carboxylic acids, amides and mineral acids to alkenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service