Documents
Z513253
Horn for 500 and 750W ultrasonic processor
High-intensity
About This Item
UNSPSC Code:
41101500
NACRES:
NB.74
material
colorless tip
manufacturer/tradename
Sonics & Materials 630-0219
General description
1/2 in. (13 mm) tip diam. For sample volumes from 10 to 250 mL.